Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcaf12l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190718.1 | NP_001177647.1 | 1 | 1027 | ||
NM_178739.6 | NP_848854.3 | 1 | 1063 | |||
GenBank mRNA | AK034343.1 | - | 1 | 880 | ||
AK034472.1 | - | 1 | 1063 | |||
AK038557.1 | - | 1 | 1018 | |||
AK039767.1 | - | 1 | 884 | |||
AK047360.1 | - | 1 | 1027 | |||
BC068319.1 | - | 1 | 867 | 30431243 |