Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178741.3 | NP_848856.1 | 10 | 2195 | ||
GenBank mRNA | AK031028.1 | - | 10 | 2196 | ||
AK034618.1 | - | 10 | 2160 | |||
AK049960.1 | - | 9 | 2083 | |||
BC086802.1 | - | 9 | 2056 | 30612176 |