Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hps5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005247.2 | NP_001005247.2 | 3 | 667 | ||
NM_001005248.2 | NP_001005248.2 | 3 | 643 | |||
NM_001167864.1 | NP_001161336.1 | 3 | 667 | |||
GenBank mRNA | AF534396.1 | - | 3 | 464 | ||
AF534397.1 | - | 3 | 440 | |||
AF534398.1 | - | 1 | 29 | |||
AF534399.1 | - | 3 | 464 | |||
AK129265.1 | - | 3 | 594 | |||
AK141088.1 | - | 3 | 465 | |||
AK154612.1 | - | 3 | 485 | |||
AK157182.1 | - | 3 | 434 | |||
BC046443.1 | - | 3 | 282 | |||
BC082542.1 | - | 3 | 667 | 6405811 |