Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdpgp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178752.3 | NP_848867.2 | 2 | 2110 | ||
GenBank mRNA | AK084531.1 | - | 2 | 2096 | ||
AK085577.1 | - | 2121 | ||||
AK086166.1 | - | 2 | 2131 | |||
AK143704.1 | - | 2 | 2136 |