Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D2hgdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178882.3 | NP_849213.2 | 10 | 2933 | ||
GenBank mRNA | AK088200.1 | - | 2879 | |||
AK088554.1 | - | 2879 |