Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Obsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178884.5 | NP_849215.3 | 4 | 1821 | ||
GenBank mRNA | AK029863.1 | - | 4 | 1611 | ||
AK031074.1 | - | 4 | 1822 | |||
AK034852.1 | - | 3 | 1883 | |||
AK048144.1 | - | 4 | 1013 | |||
AK162473.1 | - | 4 | 1731 | |||
BC051643.1 | - | 2 | 373 |