Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Garnl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178888.4 | NP_849219.2 | 4 | 308 | ||
GenBank mRNA | AK034068.1 | - | 4 | 310 | ||
AK036258.1 | - | 3 | 604 | |||
AK133163.1 | - | 5 | 482 | |||
BC057383.1 | - | 3 | 302 | |||
BC132143.1 | - | 4 | 251 | 40130420 |