Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AA792892 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178894.4 | NP_849225.2 | 3 | 571 | ||
GenBank mRNA | AK053948.1 | - | 526 | |||
AK087752.1 | - | 595 | ||||
BC120516.1 | - | 3 | 571 | 40129439 | ||
BC125434.1 | - | 3 | 571 | 40129949 |
Gene Symbol: C87414 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK136012.1 | - | 4 | 1136 | ||
BC080704.1 | - | 5 | 1284 | 30476802 | ||
BC083101.1 | - | 4 | 1135 | 30477931 |
Gene Symbol: Gm3259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270456.1 | NP_001257385.1 | 5 | 1260 | ||
GenBank mRNA | DV656723.1 | - | 1 | 57 |
Gene Symbol: Gm6367 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054302.1 | - | 1161 |