Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178726.3 | NP_848841.2 | 4 | 3376 | ||
GenBank mRNA | AK028275.1 | - | 4 | 2417 | ||
AK032529.1 | - | 4 | 3434 | |||
AK045724.1 | - | 4 | 3417 | |||
AK084918.1 | - | 1 | 348 | |||
AK131646.1 | - | 4 | 3363 | |||
AK142789.1 | - | 4 | 3396 | |||
AK147389.1 | - | 4 | 3378 | |||
AK147876.1 | - | 4 | 3414 | |||
AK165160.1 | - | 4 | 3411 | |||
AK220523.1 | - | 4 | 3386 | |||
BC096031.1 | - | 4 | 3376 | 6837066 |