Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Napepld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178728.5 | NP_848843.1 | 3, 4 | 1106 | ||
GenBank mRNA | AB112350.1 | - | 2, 3 | 949 | ||
AK041194.1 | - | 3, 4 | 1104 | |||
AK049135.1 | - | 3, 4 | 1365 | |||
AK080888.1 | - | 3, 4 | 1106 | |||
AK141585.1 | - | 3, 4 | 1386 | |||
BC062890.1 | - | 3, 4 | 1128 | 5702354 |