Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdpgp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178752.3 | NP_848867.2 | 2 | 3382 | ||
GenBank mRNA | AK085577.1 | - | 3397 | |||
BB010581.2 | - | 1 | 508 |