Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ric3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178780.3 | NP_848895.2 | 5 | 909 | ||
GenBank mRNA | AK053760.1 | - | 5 | 890 | ||
AK082275.1 | - | 5 | 890 | |||
AK149334.1 | - | 5 | 897 |