Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Obsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178884.5 | NP_849215.3 | 7 | 2780 | ||
GenBank mRNA | AK029863.1 | - | 7 | 2569 | ||
AK048144.1 | - | 7 | 1972 | |||
AK122340.1 | - | 1 | 14 | |||
AK162473.1 | - | 7 | 2690 | |||
BC051643.1 | - | 5 | 1332 | |||
BC056205.1 | - | 4 | 954 | |||
CA750706.1 | - | 3 | 699 |