Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hepacam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178899.5 | NP_849230.1 | 10 | 1992 | ||
GenBank mRNA | AK033728.1 | - | 10 | 1994 | ||
AK050501.1 | - | 10 | 1993 | |||
BC096374.1 | - | 10 | 1979 | 4483627 |