Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252458.1 | NP_001239387.1 | 19 | 3311 | ||
NM_178900.4 | NP_849231.1 | 18 | 3596 | |||
GenBank mRNA | AK037030.1 | - | 18 | 3597 |