Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277330.1 | NP_001264259.1 | 4 | 326 | ||
NM_001277805.1 | NP_001264734.1 | 4 | 324 | |||
NM_008995.2 | NP_033021.2 | 4 | 287 | |||
NM_175933.2 | NP_787947.1 | 4 | 324 | |||
GenBank mRNA | AJ416473.1 | - | 4 | 306 | ||
AK088886.1 | - | 4 | 328 | |||
AK145111.1 | - | 4 | 260 | |||
AK145361.1 | - | 4 | 261 | |||
AK161470.1 | - | 4 | 321 | |||
BC029748.1 | - | 4 | 294 | 2599159 | ||
BY122705.1 | - | 4 | 325 | |||
Z97018.1 | - | 3 | 212 |