Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281852.1 | NP_001268781.1 | 2 | 182 | ||
NM_009114.3 | NP_033140.1 | 2 | 179 | |||
GenBank mRNA | AK143826.1 | - | 2 | 153 | ||
BC027635.1 | - | 1 | 131 | 1332797 | ||
BY112406.1 | - | 2 | 149 | |||
M83219.1 | - | 2 | 149 |