Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St3gal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009179.3 | NP_033205.2 | 2 | 1360 | ||
GenBank mRNA | AK029246.1 | - | 2 | 1275 | ||
AK031052.1 | - | 2 | 1304 | |||
AK034863.1 | - | 2 | 1304 | |||
AK053827.1 | - | 2 | 1299 | |||
AK131653.1 | - | 1 | 183 | |||
BC015264.1 | - | 1 | 602 | 4211307 | ||
BC066064.1 | - | 2 | 1259 | 6850583 | ||
X76989.1 | - | 1 | 266 |