Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009720.2 | NP_033850.1 | 2, 3 | 135 | ||
GenBank mRNA | AF004591.1 | - | 2, 3 | 128 | ||
AK002432.1 | - | 2, 3 | 140 | |||
BC027632.1 | - | 2, 3 | 135 | 1328907 |