Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ghr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048178.2 | NP_001041643.1 | 2, 3 | 439 | ||
NM_001286370.1 | NP_001273299.1 | 2, 3 | 140 | |||
NM_010284.3 | NP_034414.2 | 2, 3 | 439 | |||
GenBank mRNA | AK053579.1 | - | 2, 3 | 300 | ||
AK143886.1 | - | 2, 3 | 141 | |||
AK143925.1 | - | 2, 3 | 237 | |||
AK148440.1 | - | 2, 3 | 525 | |||
BC024375.1 | - | 2, 3 | 274 | |||
BC056645.1 | - | 2, 3 | 255 | |||
BC075720.1 | - | 2, 3 | 287 | 5044060 | ||
BY754177.1 | - | 2, 3 | 440 | |||
M31680.1 | - | 2, 3 | 175 | |||
M33324.1 | - | 2, 3 | 175 |