Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013647.2 | NP_038675.2 | 3 | 303 | ||
GenBank mRNA | AK002904.1 | - | 1 | 243 | ||
AK010613.1 | - | 1 | 244 | |||
AK010641.1 | - | 1 | 244 | |||
AK011060.1 | - | 1 | 255 | |||
AK019400.2 | - | 1 | 242 | |||
AK083946.1 | - | 1 | 242 | |||
AK088413.1 | - | 1 | 243 | |||
AK088716.1 | - | 1 | 243 | |||
AK150615.1 | - | 1 | 234 | |||
AK150808.1 | - | 1 | 245 | |||
AK168207.1 | - | 1 | 242 | |||
AK169328.1 | - | 1 | 243 | |||
AV496563.1 | - | 3 | 303 | |||
BC082286.1 | - | 1 | 222 | |||
BC090618.1 | - | 1 | 270 | 5713263 | ||
BC115673.1 | - | 1 | 210 | 40104131 | ||
BC115674.1 | - | 1 | 210 | 40104132 |