Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1f6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019450.3 | NP_062323.1 | 3 | 279 | ||
GenBank mRNA | AF200493.1 | - | 2 | 118 | ||
AF206697.1 | - | 3 | 193 | |||
AK004061.1 | - | 3 | 279 | |||
AK224928.1 | - | 3 | 225 | |||
AK224929.1 | - | 3 | 225 | |||
AK224931.1 | - | 3 | 213 | |||
AK224932.1 | - | 3 | 226 | |||
BC117102.1 | - | 2 | 152 | 40126421 | ||
BC117104.1 | - | 2 | 152 | 40126423 |