Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008485.3 | NP_032511.3 | 3 | 420 | ||
GenBank mRNA | AK147105.1 | - | 3 | 421 | ||
BB617855.1 | - | 2 | 184 | |||
CJ094114.1 | - | 3 | 421 | |||
U43327.2 | - | 2 | 153 |