Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001628.2 | NP_001619.1 | 4 | 442 | ||
XM_005250234.3 | XP_005250291.1 | 4 | 435 | |||
GenBank mRNA | AB209796.1 | - | 4 | 361 | ||
AF328729.1 | - | 4 | 383 | |||
AK311339.1 | - | 4 | 465 | |||
AK313439.1 | - | 4 | 433 | |||
BC000260.1 | - | 4 | 442 | 3357652 | ||
BC005387.1 | - | 4 | 406 | 3960811 | ||
BC010391.1 | - | 4 | 408 | 4049736 | ||
BT019859.1 | - | 4 | 368 | |||
CR450351.1 | - | 4 | 368 | |||
CR542203.1 | - | 4 | 368 | |||
J04795.1 | - | 4 | 377 | |||
J05017.1 | - | 4 | 404 | |||
J05474.1 | - | 4 | 400 | |||
M34720.1 | - | 4 | 381 | |||
X15414.1 | - | 4 | 413 |