Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011213.2 | NP_035343.2 | 4, 5 | 588 | ||
GenBank mRNA | AF300943.1 | - | 2, 3 | 251 | ||
BU056582.1 | - | 4, 5 | 470 |