Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scgb1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011681.2 | NP_035811.1 | 2 | 284 | ||
GenBank mRNA | AK145844.1 | - | 2 | 249 | ||
BC027518.1 | - | 2 | 241 | 1434396 | ||
CB595042.1 | - | 2 | 288 | |||
L04503.1 | - | 2 | 310 | |||
X67702.1 | - | 2 | 291 |
Gene Symbol: Bpifb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC089529.1 | - | 1046 |