Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde6g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF190928.1 | - | 1 | 84 | ||
AK080735.1 | - | 2 | 230 | |||
BC014723.1 | - | 2 | 199 | |||
BC082288.1 | - | 2 | 187 | 6490841 | ||
Y00746.1 | - | 2 | 205 |