Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bhmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016668.3 | NP_057877.1 | 2, 3 | 432 | ||
GenBank mRNA | AF033381.1 | - | 2, 3 | 175 | ||
AK077108.1 | - | 2, 3 | 239 | |||
AK145668.1 | - | 2, 3 | 274 | |||
AK149431.1 | - | 2, 3 | 241 | |||
AK149457.1 | - | 2, 3 | 239 | |||
AK166754.1 | - | 2, 3 | 241 | |||
AK166818.1 | - | 2, 3 | 240 | |||
BC037004.1 | - | 2, 3 | 234 | 5100429 | ||
BC093510.1 | - | 2, 3 | 207 | 5101184 | ||
BC110307.1 | - | 2, 3 | 635 | 5310035 |