Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001633.3 | NP_001624.1 | 1 | 367 | ||
GenBank mRNA | AK290837.1 | - | 1 | 195 | ||
AK310776.1 | - | 1 | 294 | |||
AK310791.1 | - | 1 | 223 | |||
AY544123.1 | - | 1 | 164 | |||
BC041593.1 | - | 1 | 330 | 5433460 | ||
CB141734.1 | - | 1 | 367 | |||
X04225.1 | - | 1 | 176 | |||
X04494.1 | - | 1 | 149 |