Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dctn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159565.1 | NP_001153037.1 | 2 | 199 | ||
NM_016890.4 | NP_058586.3 | 2 | 199 | |||
GenBank mRNA | AF098508.1 | - | 2 | 178 | ||
AK002720.1 | - | 2 | 190 | |||
AK003255.1 | - | 2 | 174 | |||
AK011840.1 | - | 2 | 196 | |||
AK159344.1 | - | 2 | 197 | |||
AV493721.1 | - | 2 | 185 | |||
BC061120.1 | - | 2 | 180 | 6774742 | ||
BY141790.1 | - | 2 | 206 |