Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008073.3 | NP_032099.1 | 2 | 615 | ||
NM_177408.6 | NP_803127.3 | 2 | 615 | |||
GenBank mRNA | AF233775.1 | - | 2 | 231 | ||
AF233777.1 | - | 2 | 231 | |||
AF233782.1 | - | 2 | 231 | |||
AF233784.1 | - | 2 | 231 | |||
AF233787.1 | - | 2 | 231 | |||
AF233788.1 | - | 2 | 231 | |||
AF233789.1 | - | 2 | 231 | |||
AF233790.1 | - | 2 | 231 | |||
AF233791.1 | - | 2 | 231 | |||
AF233795.1 | - | 2 | 231 | |||
AF233798.1 | - | 2 | 231 | |||
AF233800.1 | - | 2 | 231 | |||
AF233801.1 | - | 2 | 231 | |||
AK034338.1 | - | 2 | 585 | |||
AK047085.1 | - | 2 | 193 | |||
AK049857.1 | - | 2 | 258 | |||
AK136889.1 | - | 2 | 223 | |||
AK141129.1 | - | 2 | 257 | |||
BC031762.1 | - | 2 | 433 | 4504205 | ||
M62374.1 | - | 2 | 578 |