Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008485.3 | NP_032511.3 | 17, 18 | 2762 | ||
GenBank mRNA | U43327.2 | - | 16, 17 | 2492 |