Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001635.3 | NP_001626.1 | 3 | 408 | ||
NM_139316.2 | NP_647477.1 | 3 | 408 | |||
XM_006715689.3 | XP_006715752.1 | 3 | 355 | |||
XM_006715690.3 | XP_006715753.1 | 3 | 355 | |||
XM_006715691.3 | XP_006715754.1 | 3 | 355 | |||
XM_011515271.2 | XP_011513573.1 | 3 | 355 | |||
XM_017011995.1 | XP_016867484.1 | 3 | 355 | |||
XM_017011996.1 | XP_016867485.1 | 3 | 355 | |||
GenBank mRNA | AF034996.1 | - | 3 | 261 | ||
AF498097.1 | - | 3 | 192 | |||
AK291447.1 | - | 3 | 212 | |||
BC034376.1 | - | 3 | 199 | 5181100 | ||
DA125756.1 | - | 3 | 408 | |||
U07616.1 | - | 3 | 302 | |||
X81438.1 | - | 3 | 266 |