Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009321.2 | NP_033347.1 | 4 | 291 | ||
GenBank mRNA | AK010859.1 | - | 4 | 308 | ||
AK140276.1 | - | 4 | 315 | |||
AK151218.1 | - | 4 | 292 | |||
AK168296.1 | - | 4 | 292 | |||
BC034267.1 | - | 4 | 282 | |||
BC051475.1 | - | 4 | 299 | 1430777 | ||
BC055749.1 | - | 4 | 299 | 6411373 | ||
BC087931.1 | - | 4 | 314 | 30276960 | ||
U05333.1 | - | 4 | 291 |