Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ghr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286370.1 | NP_001273299.1 | 10 | 1020 | ||
NM_010284.3 | NP_034414.2 | 10 | 1319 | |||
GenBank mRNA | AK143886.1 | - | 10 | 1021 | ||
AK143925.1 | - | 10 | 1117 | |||
BC075720.1 | - | 10 | 1167 | 5044060 | ||
M33324.1 | - | 10 | 1136 |