Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011213.2 | NP_035343.2 | 15 | 2940 | ||
GenBank mRNA | AF300943.1 | - | 13 | 2603 | ||
BC057166.1 | - | 7 | 1491 | |||
CV556730.1 | - | 3 | 345 |