Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stub1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019719.3 | NP_062693.1 | 4, 5 | 1122 | ||
GenBank mRNA | AF129086.1 | - | 4, 5 | 669 | ||
AK002752.1 | - | 4, 5 | 679 | |||
AK004464.1 | - | 4, 5 | 670 | |||
AK045776.1 | - | 4, 5 | 671 | |||
AK158595.1 | - | 4, 5 | 706 | |||
AK166630.1 | - | 4, 5 | 688 | |||
BC027427.1 | - | 4, 5 | 1122 | 4191866 | ||
BC038939.1 | - | 4, 5 | 711 | 2598940 |