Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025954.3 | NP_080230.2 | 2 | 783 | ||
GenBank mRNA | AK005906.1 | - | 2 | 455 | ||
AK010094.1 | - | 2 | 180 | |||
AK013578.1 | - | 546 | ||||
BC040100.1 | - | 2 | 783 | 4456284 | ||
BC083113.1 | - | 1 | 913 | 5356187 |