Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026610.1 | NP_080886.1 | 2, 3 | 343 | ||
GenBank mRNA | AK009807.1 | - | 2, 3 | 343 | ||
AK018707.1 | - | 2, 3 | 343 | |||
BC056229.1 | - | 2, 3 | 344 | 6475281 | ||
BC096421.1 | - | 2, 3 | 342 |