Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026614.2 | NP_080890.1 | 2, 3 | 217 | ||
GenBank mRNA | AK008155.1 | - | 2, 3 | 88 | ||
AK008156.1 | - | 2, 3 | 83 | |||
AK008286.1 | - | 2, 3 | 83 | |||
AK008414.1 | - | 2, 3 | 83 | |||
AK009338.1 | - | 2, 3 | 101 | |||
AK009396.1 | - | 2, 3 | 87 | |||
AK009461.1 | - | 2, 3 | 87 | |||
AK009493.1 | - | 2, 3 | 86 | |||
AK009608.1 | - | 2, 3 | 87 | |||
AK009642.1 | - | 2, 3 | 102 | |||
AK009652.1 | - | 2, 3 | 87 | |||
AK009775.1 | - | 2, 3 | 111 | |||
AK009840.1 | - | 2, 3 | 101 | |||
AK009985.1 | - | 2, 3 | 101 | |||
AK010024.1 | - | 2, 3 | 86 | |||
AK010090.1 | - | 2, 3 | 89 | |||
AK010092.1 | - | 2, 3 | 103 | |||
AK011579.1 | - | 2, 3 | 77 | |||
AK011947.1 | - | 2, 3 | 77 | |||
AK019084.1 | - | 2, 3 | 88 | |||
AK019201.1 | - | 2, 3 | 88 | |||
AK019207.1 | - | 2, 3 | 79 | |||
AK019215.1 | - | 2, 3 | 87 | |||
AK019242.1 | - | 2, 3 | 85 | |||
AK019245.1 | - | 2, 3 | 108 | |||
AK019291.1 | - | 2, 3 | 88 | |||
AK019301.1 | - | 2, 3 | 218 | |||
AK019324.1 | - | 2, 3 | 197 | |||
AK019354.1 | - | 2, 3 | 90 | |||
AK019359.1 | - | 2, 3 | 109 | |||
AK019364.1 | - | 2, 3 | 103 | |||
AK019368.1 | - | 2, 3 | 101 | |||
AK019430.1 | - | 2, 3 | 99 | |||
BC028633.1 | - | 2, 3 | 85 | 1247576 |