Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198831.2 | NP_942128.2 | 2, 3 | 202 | ||
GenBank mRNA | AK007653.1 | - | 2, 3 | 473 | ||
AK019161.1 | - | 2, 3 | 164 | |||
AK171917.1 | - | 2, 3 | 175 | |||
AK211110.1 | - | 1, 2 | 64 | |||
BC037190.1 | - | 2, 3 | 157 | 4168034 | ||
BC116358.1 | - | 1, 2 | 72 | 40099962 | ||
BC116359.1 | - | 1, 2 | 69 | 40099963 | ||
BC128487.1 | - | 1, 2 | 72 | 40099964 | ||
BC128488.1 | - | 1, 2 | 72 | 40099965 | ||
BE995503.1 | - | 2, 3 | 167 | |||
BG077486.2 | - | 1, 2 | 36 | |||
BU604260.1 | - | 2, 3 | 195 | |||
CD363582.1 | - | 2, 3 | 133 | |||
DN174444.1 | - | 2, 3 | 202 |