Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027445.1 | NP_081721.1 | 3, 4 | 579 | ||
GenBank mRNA | AK075579.1 | - | 3, 4 | 608 | ||
AK154071.1 | - | 3, 4 | 567 | |||
AK185969.1 | - | 1, 2 | 70 | |||
BC010777.1 | - | 3, 4 | 579 | 4241357 |