Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd33b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164441.1 | NP_001157913.1 | 1 | 489 | ||
NM_026153.3 | NP_080429.1 | 1 | 489 | |||
NM_027496.3 | NP_081772.1 | 1 | 489 | |||
GenBank mRNA | AK002572.1 | - | 1 | 284 | ||
AK012871.1 | - | 1 | 297 | |||
AK017844.1 | - | 1 | 294 | |||
AK089667.1 | - | 1 | 263 | |||
AK156757.1 | - | 1 | 457 | |||
AK170894.1 | - | 1 | 263 | |||
BC100327.1 | - | 1 | 489 |