Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srd5a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020611.4 | NP_065636.2 | 5, 6 | 782 | ||
GenBank mRNA | AK040198.1 | - | 5, 6 | 750 | ||
AK075635.1 | - | 5, 6 | 778 | |||
AK085748.1 | - | 5, 6 | 631 | |||
AK139290.1 | - | 5, 6 | 779 | |||
AK162527.1 | - | 5, 6 | 750 | |||
AK164282.1 | - | 1, 2 | 884 | |||
BC014771.1 | - | 5, 6 | 788 | 4024297 | ||
BC039564.1 | - | 5, 6 | 735 | |||
BC145647.1 | - | 5, 6 | 778 | 40130950 |