Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coprs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025556.3 | NP_079832.1 | 4 | 461 | ||
GenBank mRNA | AK008882.1 | - | 4 | 462 | ||
AK010573.1 | - | 4 | 463 | |||
BC029192.1 | - | 4 | 458 | 2647299 | ||
BC051120.1 | - | 2 | 1622 |