Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025987.3 | NP_080263.1 | 2, 3 | 335 | ||
GenBank mRNA | AK002749.1 | - | 2, 3 | 282 | ||
AK010988.1 | - | 2, 3 | 284 | |||
AK012328.1 | - | 2, 3 | 284 | |||
BC048451.1 | - | 2, 3 | 335 | 6773780 |