Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026851.2 | NP_081127.1 | 5 | 275 | ||
GenBank mRNA | AK004194.1 | - | 5 | 276 | ||
AK150806.1 | - | 5 | 275 | |||
AK151615.1 | - | 5 | 275 | |||
AK152876.1 | - | 5 | 262 | |||
AK164629.1 | - | 5 | 273 | |||
BC028522.1 | - | 5 | 308 | 1448458 | ||
BC060043.1 | - | 4 | 285 | 5683366 |