Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198831.2 | NP_942128.2 | 3, 4 | 240 | ||
GenBank mRNA | AK007653.1 | - | 3, 4 | 511 | ||
AK019161.1 | - | 3, 4 | 202 | |||
AK171917.1 | - | 3, 4 | 213 | |||
AK211110.1 | - | 2, 3 | 102 | |||
BC037190.1 | - | 3, 4 | 195 | 4168034 | ||
BC096562.1 | - | 1, 2 | 37 | |||
BC116358.1 | - | 2, 3 | 110 | 40099962 | ||
BC116359.1 | - | 2, 3 | 107 | 40099963 | ||
BC128487.1 | - | 2, 3 | 110 | 40099964 | ||
BC128488.1 | - | 2, 3 | 110 | 40099965 | ||
BG077486.2 | - | 2, 3 | 74 | |||
BU604260.1 | - | 3, 4 | 233 | |||
CD363582.1 | - | 3, 4 | 171 | |||
DN174444.1 | - | 3, 4 | 240 |