Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027088.2 | NP_081364.1 | 4 | 320 | ||
GenBank mRNA | AB047820.1 | - | 4 | 221 | ||
AK079374.1 | - | 4 | 336 | |||
AK129108.1 | - | 4 | 228 | |||
AK170576.1 | - | 4 | 321 | |||
AK171561.1 | - | 4 | 322 | |||
BC042489.1 | - | 4 | 271 | |||
BC050901.1 | - | 4 | 283 | 6402443 |