Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027445.1 | NP_081721.1 | 4, 5 | 669 | ||
GenBank mRNA | AK017523.1 | - | 5, 6 | 1105 | ||
AK030908.1 | - | 1, 2 | 1600 | |||
AK075579.1 | - | 4, 5 | 699 | |||
AK154071.1 | - | 4, 5 | 657 | |||
BC010777.1 | - | 4, 5 | 669 | 4241357 |
Gene Symbol: Syne2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005510.2 | NP_001005510.2 | 36, 37 | 5539 |